missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACCN1 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 570.00
Specifications
| Antigen | ACCN1 |
|---|---|
| Applications | Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 450 SE |
| Form | Purified |
Description
ACCN1 Polyclonal antibody specifically detects ACCN1 in Human, Rat samples. It is validated for ImmunohistochemistrySpecifications
| ACCN1 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Acid-sensing ion channel 2, Amiloride-sensitive brain sodium channel, amiloride-sensitive cation channel 1, neuronal, ASIC2a, ASIC2Mammalian degenerin homolog, BNAC1, BNaC1ACCN, BNC1Amiloride-sensitive cation channel neuronal 1, degenerin, hBNaC1, MDEGBrain sodium channel 1, neuronal amiloride-sensitive cation channel 1 | |
| CSGPGRFMAREQ (amino acid residues 15-26). | |
| Primary | |
| Store at 4°C in the dark. |
| Immunohistochemistry | |
| mFluor Violet 450 SE | |
| Rabbit | |
| Apoptosis, Cell Biology, Neuroscience | |
| 50mM Sodium Borate | |
| 40 | |
| IgG | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title