Learn More
Thermo Scientific™ Accucore™ 150-C4 HPLC Columns
Use this specially designed solid core column with a C4 functionality for biomolecules that bind too tightly to C8 or C18 columns.
Brand: Thermo Scientific™ 16526-102130
Additional Details : Weight : 0.10000kg
Beschreibung
Offer lower hydrophobic retention, ideal for optimal retention of proteins and larger peptides with Thermo Scientific™ Accucore™ 150-C4 LC Columns. Based on Core Enhanced Technology, Thermo Scientific Accucore columns provide fast, high-resolution biomolecule separations without the elevated backpressures required by sub-2μm particles.
- 150Å pore size solid core particles for fast biomoleucule separations
- Superb resolution at low backpressures
- Exceptionally rugged columns
- Significantly lower hydrophobic retention than C18
- Ideal for optimal retention of proteins and larger peptides
Ordering Information:
Other custom column dimensions are available. Please call your local Customer Service for more information.
Spezifikation
Analytical Column | |
HPLC | |
14500 psi (1000 bar) | |
2 to 9 | |
80 m²/g | |
L26 | |
Accucore | |
2% | |
Yes |
2.1 mm | |
100 mm | |
2.6 μm | |
150 Å | |
60°C | |
C4 | |
Spherical, Solid Core, Ultrapure Silica | |
Reversed Phase | |
Reversed Phase |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.