missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACD Antibody (OTI1A2), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72170APC
This item is not returnable.
View return policy
Description
ACD Monoclonal specifically detects ACD in Human samples. It is validated for Flow Cytometry.
Specifications
| ACD | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| ACD | |
| Full length recombinant protein of human ACD (NP_001075955) produced in HEK293T cells. | |
| 0.1 mL | |
| 65057.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI1A2 | |
| Flow Cytometry | |
| adrenocortical dysplasia homolog (mouse), Pip1, PIP1Tint1, POT1 and TIN2 organizing protein, POT1 and TIN2-interacting protein, Ptop, PTOPTpp1, TIN2 interacting protein 1, TINT1adrenocortical dysplasia protein homolog, TPP1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion