missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACD Antibody (OTI1A2), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72170PCP
This item is not returnable.
View return policy
Description
ACD Monoclonal specifically detects ACD in Human samples. It is validated for Flow Cytometry.
Specifications
| ACD | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| ACD | |
| Full length recombinant protein of human ACD (NP_001075955) produced in HEK293T cells. | |
| 0.1 mL | |
| 65057.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI1A2 | |
| Flow Cytometry | |
| adrenocortical dysplasia homolog (mouse), Pip1, PIP1Tint1, POT1 and TIN2 organizing protein, POT1 and TIN2-interacting protein, Ptop, PTOPTpp1, TIN2 interacting protein 1, TINT1adrenocortical dysplasia protein homolog, TPP1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction