missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACD Monoclonal Antibody (OTI2B1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525899
This item is not returnable.
View return policy
Description
ACD Monoclonal Antibody for Western Blot, IHC (P)
This gene encodes a uridine 5' -monophosphate synthase. The encoded protein is a bifunctional enzyme that catalyzes the final two steps of the de novo pyrimidine biosynthetic pathway. The first reaction is carried out by the N-terminal enzyme orotate phosphoribosyltransferase which converts orotic acid to orotidine-5' -monophosphate. The terminal reaction is carried out by the C-terminal enzyme OMP decarboxylase which converts orotidine-5' -monophosphate to uridine monophosphate. Defects in this gene are the cause of hereditary orotic aciduria. Alternate splicing results in multiple transcript variants.
Specifications
| ACD | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96AP0 | |
| ACD | |
| Full length human recombinant protein of ACD produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2B1 | |
| Unconjugated | |
| ACD | |
| Acd; ACD shelterin complex subunit and telomerase recruitment factor; ACD, shelterin complex subunit and telomerase recruitment factor; adrenocortical dysplasia; adrenocortical dysplasia homolog; adrenocortical dysplasia homolog (mouse); adrenocortical dysplasia protein; Adrenocortical dysplasia protein homolog; PIP1; POT1 and TIN2-interacting protein; PTOP; TIN2 interacting protein 1; TINT1; TPP 1; TPP I; TPP1; TPPI | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 65057 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction