missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACD Mouse anti-Human, Clone: OTI2B1, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504379S
This item is not returnable.
View return policy
Description
ACD Monoclonal antibody specifically detects ACD in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| ACD | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96AP0 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 65057 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2B1 | |
| Unconjugated | |
| ACD | |
| Acd; ACD shelterin complex subunit and telomerase recruitment factor; ACD, shelterin complex subunit and telomerase recruitment factor; adrenocortical dysplasia; adrenocortical dysplasia homolog; adrenocortical dysplasia homolog (mouse); adrenocortical dysplasia protein; Adrenocortical dysplasia protein homolog; PIP1; POT1 and TIN2-interacting protein; PTOP; TIN2 interacting protein 1; TINT1; TPP 1; TPP I; TPP1; TPPI | |
| Full length human recombinant protein of human ACD produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction