missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACE-2 Antibody (2818I), HRP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB34372H
This item is not returnable.
View return policy
Description
ACE-2 Monoclonal antibody specifically detects ACE-2 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry
Specifications
| ACE-2 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, Flow Cytometry, Immunocytochemistry | |
| 2818I | |
| Western Blot, Flow Cytometry, Immunocytochemistry | |
| ACEHangiotensin I converting enzyme 2, ACE-related carboxypeptidase, angiotensin I converting enzyme (peptidyl-dipeptidase A) 2, angiotensin-converting enzyme 2, Angiotensin-converting enzyme homolog, DKFZp434A014, EC 3.4.17, EC 3.4.17.23, Metalloprotease MPROT15 | |
| Chinese Hamster Ovary cell line CHO-derived mouse ACE-2, Gln18-Thr740, Accession # NP_081562 | |
| 0.1 mL | |
| Immunology, Virology Bacteria and Parasites | |
| 59272 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction