Learn More
ACE-2 Antibody (535919) [DyLight 755], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB9332Z
Product Code. 18519609
  • € 652.00 / 0.10mL
Define Immunology Research on Your Terms With R&D Systems
Use promo code 24090
View all promotions
Description
ACE-2 Monoclonal antibody specifically detects ACE-2 in Human, Rat, Hamster samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ACE-2
Monoclonal
DyLight 755
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human, Rat, Hamster
Purified
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF
535919
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready
ACEHangiotensin I converting enzyme 2, ACE-related carboxypeptidase, angiotensin I converting enzyme (peptidyl-dipeptidase A) 2, angiotensin-converting enzyme 2, Angiotensin-converting enzyme homolog, DKFZp434A014, EC 3.4.17, EC 3.4.17.23, Metalloprotease MPROT15
Mouse myeloma cell line NS0-derived recombinant human ACE-2, Gln18-Ser740, Accession # Q9BYF1
0.1 mL
Immunology, Virology Bacteria and Parasites
59272
Store at 4°C in the dark.
IgG2a