missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACE/CD143 Antibody (230214) [FITC], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB15131F
This item is not returnable.
View return policy
Description
ACE/CD143 Monoclonal antibody specifically detects ACE/CD143 in Mouse samples. It is validated for Flow Cytometry, Immunoprecipitation, CyTOF
Specifications
| ACE/CD143 | |
| Monoclonal | |
| FITC | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, Immunoprecipitation, CyTOF | |
| 230214 | |
| Flow Cytometry, Immunoprecipitation, CyTOF-ready | |
| ACE1angiotensin converting enzyme, somatic isoform, angiotensin I converting enzyme (peptidyl-dipeptidase A) 1, carboxycathepsin, CD143, CD143 antigen, DCP, DCP1angiotensin-converting enzyme, dipeptidyl carboxypeptidase 1, Dipeptidyl carboxypeptidase I, EC 3.2.1.-, EC 3.4.15.1, Kininase II, MGC26566, MVCD3, peptidase P, testicular ECA | |
| Mouse myeloma cell line NS0-derived recombinant mouse ACE/CD143, Leu35-Glu1264, Accession # P09470 | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, CD Markers, Cell Biology, Endocrinology, Immunology, Signal Transduction, Stem Cells | |
| 1636 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction