missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACE/CD143 Antibody (ACE/3763), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08558AF700
This item is not returnable.
View return policy
Description
ACE/CD143 Monoclonal antibody specifically detects ACE/CD143 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin)
Specifications
| ACE/CD143 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| ACE1angiotensin converting enzyme, somatic isoform, angiotensin I converting enzyme (peptidyl-dipeptidase A) 1, carboxycathepsin, CD143, CD143 antigen, DCP, DCP1angiotensin-converting enzyme, dipeptidyl carboxypeptidase 1, Dipeptidyl carboxypeptidase I, EC 3.2.1.-, EC 3.4.15.1, Kininase II, MGC26566, MVCD3, peptidase P, testicular ECA | |
| Recombinant fragment (around aa 652-788) of human ACE/CD143 protein (Uniprot: P12821) | |
| 100 μg | |
| Cancer, Cardiovascular Biology, CD Markers, Cell Biology, Endocrinology, Immunology, Signal Transduction, Stem Cells | |
| 1636 | |
| Store at 4°C in the dark. | |
| IgG2c κ |
| Western Blot, ELISA, Immunohistochemistry (Paraffin) | |
| ACE/3763 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction