missing translation for 'onlineSavingsMsg'
Learn More

ACE/CD143 Antibody (S01-9E3), Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NBP3-19570-100ul

 View more versions of this product

Product Code. 18690774

  • € 645.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ACE/CD143 Monoclonal antibody specifically detects ACE/CD143 in Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ACE/CD143
Monoclonal
0.3 mg/mL
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
Primary
Mouse, Rat
Purified
Western Blot
S01-9E3
Unconjugated
ACE1angiotensin converting enzyme, somatic isoform, angiotensin I converting enzyme (peptidyl-dipeptidase A) 1, carboxycathepsin, CD143, CD143 antigen, DCP, DCP1angiotensin-converting enzyme, dipeptidyl carboxypeptidase 1, Dipeptidyl carboxypeptidase I, EC 3.2.1.-, EC 3.4.15.1, Kininase II, MGC26566, MVCD3, peptidase P, testicular ECA
Recombinant protein of mouse ACE/CD143 (Uniprot # P09470)
100 μg
Cancer, Cardiovascular Biology, CD Markers, Cell Biology, Endocrinology, Immunology, Signal Transduction, Stem Cells
1636
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.