missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACE2 Mouse anti-Human, Clone: OTI2G7, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803845S
This item is not returnable.
View return policy
Description
ACE2 Monoclonal antibody specifically detects ACE2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| ACE2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BYF1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 59272 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2G7 | |
| Unconjugated | |
| ACE2 | |
| 2010305L05Rik; ACE 2; Ace2; ACEH; ACE-related carboxypeptidase; angiotensin I converting enzyme (peptidyl-dipeptidase A) 2; angiotensin I converting enzyme 2; angiotensin-converting enzyme 2; Angiotensin-converting enzyme homolog; anigotensin I converting enzyme 2; anigotensin-converting enzyme-related carboxypeptidase; metalloprotease MPROT15; peptidyl-dipeptidase A; Processed angiotensin-converting enzyme 2; renal angiotensin-converting enzyme 2; UNQ868/PRO1885 | |
| Human recombinant protein fragment corresponding to amino acids 18-237 of human ACE2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction