missing translation for 'onlineSavingsMsg'
Learn More

Aceclofenac EP Impurity D, Mikromol™

Product Code. 18030480
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18030480 25 mg 25mg
18001680 100 mg 100mg
30459401 4 x 25 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18030480 Supplier Mikromol Supplier No. MM0181.010025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Aceclofenac EP Impurity D
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aceclofenac Methyl Ester
CAS 139272-66-5
Analyte or Component Names Aceclofenac Methyl Ester
Shipping Condition Room Temperature
Molecular Formula C17 H15 Cl2 N O4
Certifications/Compliance ISO 17025
Synonym Methyl [[[2-[(2,6-Dichlorophenyl)amino]phenyl]acetyl]oxy]acetate (Methyl Ester of Aceclofenac), Methyl [[[2-[(2,6-Dichlorophenyl)amino]phenyl]acetyl]oxy]acetate, Aceclofenac Imp. D (EP), Methyl Ester of Aceclofenac, Aceclofenac Impurity D
InChI Formula InChI=1S/C17H15Cl2NO4/c1-23-16(22)10-24-15(21)9-11-5-2-3-8-14(11)20-17-12(18)6-4-7-13(17)19/h2-8,20H,9-10H2,1H3
SMILES COC(=O)COC(=O)Cc1ccccc1Nc2c(Cl)cccc2Cl
IUPAC Name (2-methoxy-2-oxoethyl) 2-[2-(2,6-dichloroanilino)phenyl]acetate
Molecular Weight (g/mol) 368.21
Quantity 25 mg
Formula Weight 367.0378 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.