Learn More
MedChemExpress Acelarin
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Molecular Weight (g/mol) | 580.47 g/mol |
| Product Type | Reference compound |
| Content And Storage | Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month |
| Shipping Condition | Room temperature |
| Purity | 98.1% |
| Quantity | 5 mg |
| Solubility Information | DMSO : ≥ 36 mg/mL (62.02 mM) |
| For Use With (Application) | Cancer-Kinase/protease |
| CAS | 840506-29-8 |
| Synonym | NUC-1031 |