missing translation for 'onlineSavingsMsg'
Learn More

Acetaldehyde Ammonia Trimer, TRC

Product Code. 17972945
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17972945 5 g 5g
30133196 25 g 25g
17922075 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17972945 Supplier Toronto Research Chemicals Supplier No. TRCA1327435G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58052-80-5
InChI Formula InChI=1S/C6H15N3.3H2O/c1-4-7-5(2)9-6(3)8-4;;;/h4-9H,1-3H3;3*1H2
IUPAC Name 2,4,6-trimethyl-1,3,5-triazinane;trihydrate
Molecular Formula C6H21N3O3
Molecular Weight (g/mol) 183.25
SMILES CC(NC(C)N1)NC1C.[3H2O]
Synonym Hexahydro-2,4,6-trimethyl s-Triazine Trihydrate, Hexahydro-2,4,6-trimethyl-1,3,5-triazine Trihydrate
Chemical Name or Material Acetaldehyde Ammonia Trimer

Specifications

Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.