missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, TRC

Product Code. 30003600
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
Unit Size:
1 L
2.5 L
5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003600 5 L 5 L
30008552 2.5 L 2.5 L
17943705 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003600 Supplier Toronto Research Chemicals Supplier No. TRCA1676405L

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2 H4 O2
Molecular Weight (g/mol) 60.05
Synonym PN: WO2017048620 SEQID: 12 claimed sequence, Aci-Jel, E 260, Engage Agro, Ethanoic acid, Ethanoic acid monomer, Ethylic acid, Glacial acetic acid, HACP 85, Methanecarboxylic acid, Munger Horticultural Vinegar Plus, NSC 111201, NSC 112209, NSC 115870, NSC 127175, NSC 132953, NSC 406306, Vinegar acid, Weed Pharm, acetic acid
IUPAC Name acetic acid
SMILES CC(=O)O

Specifications

InChI Formula InChI=1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)
Formula Weight 60.0211
Quantity 5 L
Chemical Name or Material Acetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.