missing translation for 'onlineSavingsMsg'
Learn More

Acetoacetyl Coenzyme A Sodium Hydrate (>90%), TRC

Product Code. 30312979
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
1mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30312979 25 mg 25mg
30318491 1 mg 1mg
30310719 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30312979 Supplier Toronto Research Chemicals Supplier No. TRCA16184025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acetoacetyl Coenzyme A Sodium Salt Hydrate
Analyte or Component Names Acetoacetyl Coenzyme A Sodium Salt Hydrate
Shipping Condition Blue Ice Dry Pack
Molecular Formula C25 H40 N7 O18 P3 S . xNa . xH2O
InChI Formula InChI=1S/C25H40N7O18P3S.Na.H2O/c1-13(33)8-16(35)54-7-6-27-15(34)4-5-28-23(38)20(37)25(2,3)10-47-53(44,45)50-52(42,43)46-9-14-19(49-51(39,40)41)18(36)24(48-14)32-12-31-17-21(26)29-11-30-22(17)32;;/h11-12,14,18-20,24,36-37H,4-10H2,1-3H3,(H,27,34)(H,28,38)(H,42,43)(H,44,45)(H2,26,29,30)(H2,39,40,41);;1H2/q;+1;/t14-,18-,19-,20+,24-;;/m1../s1
Solution Type Neat
SMILES O.[Na+].CC(=O)CC(=O)SCCNC(=O)CCNC(=O)[C@H](O)C(C)(C)COP(=O)(O)OP(=O)(O)OC[C@H]1O[C@H]([C@H](O)[C@@H]1OP(=O)(O)O)n2cnc3c(N)ncnc23
IUPAC Name sodium;S-[2-[3-[[(2R)-4-[[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-4-hydroxy-3-phosphonooxy-tetrahydrofuran-2-yl]methoxy-hydroxy-phosphoryl]oxy-hydroxy-phosphoryl]oxy-2-hydroxy-3,3-dimethyl-butanoyl]amino]propanoylamino]ethyl] 3-oxobutanethioate;hydrate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 892.6
Quantity 25 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.