Learn More
Acetone 2,4-Dinitrophenylhydrazone, TRC
Specifications
Specifications
| Chemical Name or Material | Acetone-DNPH |
| CAS | 1567-89-1 |
| Molecular Formula | C9 H10 N4 O4 |
| Quantity | 1 g |
| Synonym | 2-Propanone, (2,4-dinitrophenyl)hydrazone (9CI), Acetone, (2,4-dinitrophenyl)hydrazone (6CI,8CI), 1-(2,4-Dinitrophenyl)-2-(propan-2-ylidene)hydrazine, NSC 131450, NSC 6120 |
| InChI Formula | InChI=1S/C9H10N4O4/c1-6(2)10-11-8-4-3-7(12(14)15)5-9(8)13(16)17/h3-5,11H,1-2H3 |
| SMILES | CC(=NNc1ccc(cc1[N+](=O)[O-])[N+](=O)[O-])C |
| IUPAC Name | 2,4-dinitro-N-(propan-2-ylideneamino)aniline |
| Molecular Weight (g/mol) | 238.2 |
| Formula Weight | 238.0702 |