missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetyl-CoA Carboxylase alpha/ACACA [p Ser79] Antibody (JE55-54), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33134
This item is not returnable.
View return policy
Description
Acetyl-CoA Carboxylase alpha/ACACA Monoclonal antibody specifically detects Acetyl-CoA Carboxylase alpha/ACACA in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| Acetyl-CoA Carboxylase alpha/ACACA | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000 | |
| ACACacetyl-CoA carboxylase 1, ACC1ACC, ACCA, ACC-alpha, acetyl-CoA carboxylase alpha, acetyl-Coenzyme A carboxylase alpha, EC 6.4.1.2 | |
| Synthetic phosphopeptide corresponding to residues surrounding Ser79 of human Acetyl-CoA Carboxylase alpha/ACACA protein. (Uniprot: Q13085) | |
| 100 μL | |
| Breast Cancer, Lipid and Metabolism, Neuroscience, Phospho Specific | |
| 31 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| JE55-54 | |
| Unconjugated | |
| 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction