missing translation for 'onlineSavingsMsg'
Learn More

Acetyl-CoA Carboxylase [p Ser79] Antibody, Novus Biologicals™

Product Code. 30226787 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30226787 20 μL 20µL
30229803 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30226787 Supplier Novus Biologicals Supplier No. NBP33357320ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Acetyl-CoA Carboxylase Monoclonal antibody specifically detects Acetyl-CoA Carboxylase in Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Acetyl-CoA Carboxylase
Applications ELISA, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Dilution Western Blot 1:2000 - 1:8000, ELISA Recommended starting concentration is 1 μg/mL
Formulation PBS (pH 7.3), 50% glycerol, 0.05% BSA
Gene Alias ACAC, Acac1, ACACAD, ACC, ACC1, ACCA, ACC-alpha, ACCalphaACACalpha, acetyl-CoA carboxylase 1, acetyl-Coenzyme A carboxylase alpha, hACC1
Host Species Rabbit
Immunogen A synthetic phosphorylated peptide around S79 of human Acetyl-CoA Carboxylase
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 31
Target Species Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.