missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetyl-CoA Carboxylase [p Ser79] Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33573-100ul
This item is not returnable.
View return policy
Description
Acetyl-CoA Carboxylase Monoclonal antibody specifically detects Acetyl-CoA Carboxylase in Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| Acetyl-CoA Carboxylase | |
| Monoclonal | |
| Western Blot 1:2000 - 1:8000, ELISA Recommended starting concentration is 1 μg/mL | |
| ACAC, Acac1, ACACAD, ACC, ACC1, ACCA, ACC-alpha, ACCalphaACACalpha, acetyl-CoA carboxylase 1, acetyl-Coenzyme A carboxylase alpha, hACC1 | |
| A synthetic phosphorylated peptide around S79 of human Acetyl-CoA Carboxylase | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 31 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction