missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetylated Lysine Mouse anti-Mouse, Biotin, Clone:7F8, Abnova™
Mouse monoclonal antibody raised against Acetylated Lysine .
Brand: Abnova MAB16766.100ug
This item is not returnable.
View return policy
Specifications
| Acetylated Lysine | |
| Monoclonal | |
| Biotin | |
| In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide) | |
| Acetylated KLH. | |
| Mouse | |
| Antibody | |
| IgG1 |
| ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 7F8 | |
| ELISA Immunocytochemistry Immunofluorescence Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100) Immunoprecipitation Western Blot (1:1000) The optimal working dilution should be determined by the end user. |
|
| Mouse | |
| 100 μg | |
| Store at -20°C. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction