missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetylcholine Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-64656AF350
This item is not returnable.
View return policy
Description
Acetylcholine Polyclonal antibody specifically detects Acetylcholine in All samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)
Specifications
| Acetylcholine | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| All species | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| Alexa Fluor 350 | |
| [2 (acetyloxy)ethyl]trimethylammonium, ACh | |
| Synthetic acetylcholine conjugated to Keyhole limpet Haemocyanin. | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction