missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetylcholinesterase/ACHE Antibody (HR2), DyLight 650 , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB300-528C
This item is not returnable.
View return policy
Description
Acetylcholinesterase/ACHE Monoclonal antibody specifically detects Acetylcholinesterase/ACHE in Human, Bovine, Feline, Rabbit, Mouse (Negative), Rat (Negative) samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifications
| Acetylcholinesterase/ACHE | |
| Monoclonal | |
| DyLight 650 | |
| acetylcholinesterase, acetylcholinesterase (Yt blood group), AChE, apoptosis-related acetylcholinesterase, ARACHE, EC 3.1.1, EC 3.1.1.7, N-ACHE, YT, Yt blood group | |
| Purified human cerebellar acetylcholinesterase. | |
| 0.2 mL | |
| Cellular Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 43 | |
| Store at 4°C in the dark. | |
| IgG2b |
| ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| HR2 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Bovine, Feline, Rabbit, Mouse (Negative) | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction