missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acid Number (AN) Standard: 3.0 mg KOH/g in Hydrocarbon Oil, VHG™
Click to view available options
Quantity:
50 g
400 g
800 g
Unit Size:
400g
50g
800g
Description
Acid Number (AN) Standard: 3.0 mg KOH/g in Hydrocarbon Oil
Specifications
Specifications
| Chemical Name or Material | Potassium hydroxide |
| CAS | 1310-58-3 |
| Methods | Potentiometric Titration |
| Analyte or Component Names | Potassium hydroxide |
| Shipping Condition | Room Temperature |
| Molecular Formula | H K O |
| Concentration | 3 mg/g |
| Certifications/Compliance | ISO 17034 |
| Synonym | Potassium hydroxide |
| InChI Formula | InChI=1S/K.H2O/h;1H2/q+1;/p-1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction