missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Acid Phosphatase 2 Polyclonal Antibody, Invitrogen™
GREENER_CHOICE

Product Code. p-200020184
Change view
Click to view available options
Quantity:
20 μL
200 μL
Unit Size:
200µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17929822 200 μL 200µL
17900203 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17929822 Supplier Invitrogen™ Supplier No. PA5145477

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This gene encodes the beta subunit of lysosomal acid phosphatase (LAP). LAP is chemically and genetically distinct from red cell acid phosphatase. The encoded protein belongs to a family of distinct isoenzymes which hydrolyze orthophosphoric monoesters to alcohol and phosphate. Mutations in this gene or in the related alpha subunit gene cause acid phosphatase deficiency. Multiple alternatively spliced transcript variants encoding different isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Acid Phosphatase 2
Applications Western Blot, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene Acp2
Gene Accession No. P11117, P20611, P24638
Gene Alias acid phosphatase 2, lysosomal; Acid phosphatase 2, lysozymal; Acp2; Acp-2; LAP; LMW-PTP-II low molecular weight phosphotyrosine protein phosphatase isozyme 2; lysosomal acid phosphatase; Unknown (protein for MGC:142461)
Gene Symbols Acp2
Host Species Rabbit
Immunogen Recombinant Human ACP2 protein, Tyr40-Ala250 (Accession #P11117).
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11432, 24162, 53
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.