missing translation for 'onlineSavingsMsg'
Learn More

Acid Red 88 (Technical Grade), TRC

Product Code. 30348556
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30348556 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30348556 Supplier Toronto Research Chemicals Supplier No. TRCA795243100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acid Red 88 (Technical Grade)
CAS 1658-56-6
Molecular Formula C20H13N2NaO4S
Quantity 100 g
Synonym C.I. Acid Red 88, Monosodium Salt, Naphthalene Red J, 11391 Red, 2-Naphthol Red J, Acid Cardinal G, Acid Fast Red A, Acid Leather Red ROC, Acid Red A, Acid Red A (Chinese), Acid Red AV, Acid Red G, Acid Rose AV, Acid Scarlet G, Airedale Red A, Amacid Fast Red A, Ambicid Fast Red E, Anadurm Red A-ROC, Anthrosin BRX, Apollo Acid Rocceline, Atul Acid Fast Red A, Azo Acid Red GS, Basacid Red 340, Benzyl Red ROC, Benzyl Red S, Brasilan Red S, Bucacid Fast Red A, C.I. 15620, C.I. Acid Red 88, Calcocid Fast Red A, Cavalene Red A, Colacid Red AV, Colocid Fast Red A, Conacid Red MM, Daedo Acid Roccelline NS, Dai-ei Roccelline, Derma Fur Red R 150, Diacid Red A, Dinacid Fast Red A, Dyacid Red J, Dycosacid Red A, Eniacid Fast Red A, Eriosin Roccelline, Eriosin Roccelline SS, Ext D and C Red No. 8Fabracid Red S-A, Fast Acid Red G, Fast Red A, Fast Red A (acid dye), Fast Red AE, Fast Red AG, Fast Red ALS, Fast Red AV, Fast Red MA, Fast Red S, Fenazo Red M, Fur Red KB, Hidacid Fast Red A, Hispacid Fast Red A, Ichoacid Red A, Indacid Acid Red A, Japan Red 506, Japan Red No. 506, Kayaku Roccelline, Kemacid Fast Red A, Libacid Fast Red A, Lurazol Red E, Monacid Red A, Multacid Red A, Multicuer Red A, Naphthalene Red JS, Naphtocard Fast Red C, Neklacid Fast Red A, Neutral Red R, New Red WO, Nitto Roccelline, Peony, Plastoresin Red RC, Pontacyl Fast Red AS, Red J, Red No. 506, Red for Furs G, Rifa Acid Roccelline NS, Rifa Leather Red E, Ritacid Rosalin AV, Rocceline, Roccelline, Roccelline A, Roccelline B, Roccelline G, Roccelline K, Roccelline KG, Roccelline L, Roccelline NS, Roccelline NS 200, Roccelline S, Rocelline, Romexal Red AV, Sandal Acid Red G, Shikiso Roccelline, Sodium 4-[(2-hydroxy-1-naphthyl)azo]-1-naphthalene Sulfonate, Tertracid Red RO, Toyo Roccelline, Triacid Fast Red Av, Triacid Red AN, Vicoacid Fast Red A, Vondacid Red GN, Vopsider Red AAV, 4-[(2-Hydroxy-1-naphthalenyl)azo]-1-naphthalenesulfonic Acid Monosodium Salt
InChI Formula InChI=1S/C20H14N2O4S.Na/c23-18-11-9-13-5-1-2-6-14(13)20(18)22-21-17-10-12-19(27(24,25)26)16-8-4-3-7-15(16)17;/h1-12,23H,(H,24,25,26);/q;+1/p-1/b22-21+;
SMILES [Na+].Oc1ccc2ccccc2c1N=Nc3ccc(c4ccccc34)S(=O)(=O)[O-]
IUPAC Name sodium;4-[(E)-(2-hydroxynaphthalen-1-yl)diazenyl]naphthalene-1-sulfonate
Molecular Weight (g/mol) 400.38
Formula Weight 400.0494
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.