missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acinus Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB500-217IR
This item is not returnable.
View return policy
Description
Acinus Polyclonal antibody specifically detects Acinus in Human samples. It is validated for Western Blot
Specifications
| Acinus | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 755 | |
| Acinus, ACN, apoptotic chromatin condensation inducer 1, apoptotic chromatin condensation inducer in the nucleus, fSAP152, functional spliceosome-associated protein 152, KIAA0670DKFZp667N107 | |
| A peptide made to the C-terminal sequence RNRQLEREKRREHS. | |
| 0.1 mL | |
| Apoptosis, Cancer, Chromatin Research, Tumor Suppressors | |
| 22985 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction