missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACK1 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97358DL594
This item is not returnable.
View return policy
Description
ACK1 Polyclonal antibody specifically detects ACK1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| ACK1 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| DyLight 594 | |
| ACK-1, ACK1ACKp21cdc42Hs, activated CDC42 kinase 1, activated Cdc42-associated kinase 1, activated p21cdc42Hs kinase, EC 2.7.10, EC 2.7.10.2, FLJ44758, FLJ45547, Tyrosine kinase non-receptor protein 2, tyrosine kinase, non-receptor, 2 | |
| Produced in rabbits immunized with E. coli-derived Human ACK1 fragment. | |
| 0.1 mL | |
| Protein Kinase, Signal Transduction | |
| 10188 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction