missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACMSD Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | ACMSD |
|---|---|
| Concentration | 400 μg/mL |
| Applications | Immunohistochemistry, ELISA |
| Form | Liquid |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18056195
|
Proteintech
26834-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18080616
|
Proteintech
26834-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The ACMSD antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ACMSD. This antibody recognizes human, mouse antigen. The ACMSD antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
| ACMSD | |
| Immunohistochemistry, ELISA | |
| Rabbit | |
| 130013 | |
| IgG | |
| Store at -20°C. Stable for one year after shipment. |
| 400 μg/mL | |
| Liquid | |
| Human, Mouse | |
| ACMSD, Picolinate carboxylase | |
| Primary |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit