missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SDHAF3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13272169
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13272169 400 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13272169 Supplier Invitrogen™ Supplier No. PA524526

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SDHAF3 Polyclonal Antibody for Western Blot, IHC (P)

Plays an essential role in the assembly of succinate dehydrogenase (SDH), an enzyme complex (also referred to as respiratory complex II) that is a component of both the tricarboxylic acid (TCA) cycle and the mitochondrial electron transport chain, and which couples the oxidation of succinate to fumarate with the reduction of ubiquinone (coenzyme Q) to ubiquinol. Promotes maturation of the iron-sulfur protein subunit SDHB of the SDH catalytic dimer, protecting it from the deleterious effects of oxidants. May act together with SDHAF1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SDHAF3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene SDHAF3
Gene Accession No. Q8BQU3, Q9NRP4
Gene Alias 0610005M07Rik; 4933430A16Rik; Acn9; ACN9 homolog; ACN9 homolog (S. cerevisiae); DC11; Liver regeneration-related protein LRRGT00092; LRRGT00092; LYRM10; protein ACN9 homolog, mitochondrial; SDH assembly factor 3; Sdh7; SDHAF3; Succinate dehydrogenase assembly factor 3, mitochondrial; succinate dehydrogenase complex assembly factor 3
Gene Symbols SDHAF3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 13-42 amino acids from the N-terminal region of human ACN9.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57001, 71238
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.