missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aconitase 2 Antibody (OTI3G8), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70074FR
This item is not returnable.
View return policy
Description
Aconitase 2 Monoclonal specifically detects Aconitase 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Aconitase 2 | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| ACO2 | |
| Full length recombinant protein of human Aconitase 2 (NP_001089) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI3G8 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Aconitase, aconitase 2, mitochondrial, aconitate hydratase, mitochondrial, ACONM, Citrate hydro-lyase, EC 4.2.1, EC 4.2.1.3, MGC20605, MGC33908 | |
| Mouse | |
| Affinity Purified | |
| Lipid and Metabolism | |
| 50.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction