missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aconitate hydratase B Antibody (09), Novus Biologicals™
Mouse Monoclonal Antibody
€ 220.00 - € 362.00
Specifications
| Antigen | Aconitate hydratase B |
|---|---|
| Clone | 09 |
| Dilution | Sandwich ELISA Capture 1:250-1:2000 |
| Applications | Sandwich ELISA Capture |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18636506
|
Novus Biologicals
NBP3-06091-100ul |
100 μg |
€ 362.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18699048
|
Novus Biologicals
NBP3-06091-50ul |
50 μg |
€ 220.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Aconitate hydratase B Monoclonal antibody specifically detects Aconitate hydratase B in E. coli samples. It is validated for Sandwich ELISA CaptureSpecifications
| Aconitate hydratase B | |
| Sandwich ELISA Capture 1:250-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Aconitate hydratase B (Uniprot#: P36683; Met1-Val865). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 09 | |
| Sandwich ELISA Capture | |
| Unconjugated | |
| Mouse | |
| E. coli | |
| acnB, hypothetical protein, yacI, yacJ | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title