Learn More
Aconitate hydratase B Antibody (102), Janelia Fluor™ 646, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-90177JF646
Product Code. 18485459
  • € 607.00 / 0.10mL
Description
Aconitate hydratase B Monoclonal antibody specifically detects Aconitate hydratase B in E. coli samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
102
Rabbit
Purified
Janelia Fluor 646
Store at 4C in the dark.