missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aconitate hydratase B Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06108MFV500
This item is not returnable.
View return policy
Description
Aconitate hydratase B Polyclonal antibody specifically detects Aconitate hydratase B in E. coli samples. It is validated for ELISA
Specifications
| Aconitate hydratase B | |
| Polyclonal | |
| ELISA | |
| acnB, hypothetical protein, yacI, yacJ | |
| Produced in rabbits immunized with purified, recombinant Aconitate hydratase B (Uniprot#: P36683; Met 1-Val 865). | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG |
| ELISA | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| E. coli | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction