missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACOT12 Monoclonal Antibody (OTI1D3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502530S
This item is not returnable.
View return policy
Description
ACOT12 Monoclonal antibody specifically detects ACOT12 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| ACOT12 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BQ13 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 65987 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Flow Cytometry, Western Blot | |
| OTI1D3 | |
| Unconjugated | |
| KCTD14 | |
| 1300004O04Rik; 4930449F15Rik; Acetyl-coenzyme A thioesterase; Acot12; Acyl-CoA thioester hydrolase 12; acyl-CoA thioesterase 12; Acyl-coenzyme A thioesterase 12; AV027244; Cach; CACH1; CACH-1; Cytoplasmic acetyl-CoA hydrolase 1; cytosolic acetyl-CoA hydrolase; hCACH-1; mCACH-1; rACH; rCACH-1; STARD15; StAR-related lipid transfer (START) domain containing 15; START domain-containing protein 15; THEAL | |
| Full length human recombinant protein of human KCTD14 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction