missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACOT12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531981
This item is not returnable.
View return policy
Description
Recommended positive controls: A549, HeLa. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Hydrolyzes acetyl-CoA to acetate and CoA.
Specifications
| ACOT12 | |
| Polyclonal | |
| Unconjugated | |
| ACOT12 | |
| 1300004O04Rik; 4930449F15Rik; Acetyl-coenzyme A thioesterase; Acot12; Acyl-CoA thioester hydrolase 12; acyl-CoA thioesterase 12; Acyl-coenzyme A thioesterase 12; AV027244; Cach; CACH1; CACH-1; Cytoplasmic acetyl-CoA hydrolase 1; cytosolic acetyl-CoA hydrolase; hCACH-1; mCACH-1; rACH; rCACH-1; STARD15; StAR-related lipid transfer (START) domain containing 15; START domain-containing protein 15; THEAL | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 134526 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q8WYK0 | |
| ACOT12 | |
| Recombinant protein encompassing a sequence within the center region of human ACOT12. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction