missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ACOT12 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15693895
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15693895 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15693895 Supplier Invitrogen™ Supplier No. PA549968

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total ACOT12 protein.

Hydrolyzes acetyl-CoA to acetate and CoA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACOT12
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ACOT12
Gene Accession No. Q8WYK0
Gene Alias 1300004O04Rik; 4930449F15Rik; Acetyl-coenzyme A thioesterase; Acot12; Acyl-CoA thioester hydrolase 12; acyl-CoA thioesterase 12; Acyl-coenzyme A thioesterase 12; AV027244; Cach; CACH1; CACH-1; Cytoplasmic acetyl-CoA hydrolase 1; cytosolic acetyl-CoA hydrolase; hCACH-1; mCACH-1; rACH; rCACH-1; STARD15; StAR-related lipid transfer (START) domain containing 15; START domain-containing protein 15; THEAL
Gene Symbols ACOT12
Host Species Rabbit
Immunogen Synthesized peptide derived from internal of human ACOT12 (aa 281-330).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 134526
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.