missing translation for 'onlineSavingsMsg'
Learn More

ACOT2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

€ 280.00 - € 624.00

Specifications

Antigen ACOT2
Dilution Western Blot 0.04-0.4 μg/ml, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18637346
View Documents
Novus Biologicals
NBP2-62632-25ul
25 μL
€ 280.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18631728
View Documents
Novus Biologicals
NBP2-62632
100 μg
€ 624.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ACOT2 Polyclonal antibody specifically detects ACOT2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

ACOT2
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction
PBS (pH 7.2) and 40% Glycerol
10965
IgG
Protein A purified
Western Blot 0.04-0.4 μg/ml, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000
Polyclonal
Purified
RUO
Human
acyl-CoA thioesterase 2MTE1, Acyl-coenzyme A thioester hydrolase 2a, acyl-coenzyme A thioesterase 2, mitochondrial, CTE-Ia, EC 3.1.2.2, Long-chain acyl-CoA thioesterase 2, mitochondrial acyl-CoA thioesterase 1, mitochondrial acyl-CoA thioesterase 2, peroxisomal long-chain acyl-coA thioesterase 2, PTE2A, PTE2CTE1A, ZAP128Mte1
This antibody was developed against a recombinant protein corresponding to amino acids: MSNKLLSPHPHSVVLRSEFKMASSPAVLRASRLYQWSLKSSAQFLGSPQLRQVGQIIRVPAR
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.