Learn More
ACOT2 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-15633-100UL
 View more versions of this product
Product Code. 18047095
  • € 450.00 / 100µL
Description
The ACOT2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ACOT2. This antibody recognizes human, mouse, rat antigen. The ACOT2 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
ACOT2
1000 μg/mL
ACOT2, acyl CoA thioesterase 2, CTE Ia, Mte1, PTE2, PTE2A, ZAP128
100 μL
10965
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Immunofluorescence, Flow Cytometry (Intracellular Staining)
CoraLite Plus 488
Rabbit
Primary
Human, Mouse, Rat
150 kDa