missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACOT6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-31708-25ul
This item is not returnable.
View return policy
Description
ACOT6 Polyclonal specifically detects ACOT6 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| ACOT6 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Q3I5F7 | |
| ACOT6 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: ATVLINACVANTVAPLHYKDMIIPKLVDDLGKVKITKSGFLTFMDTWSNPLEEHNHQSLVPLEKAQVPFLFIVGMDDQSWKSEFYAQI | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| acyl-CoA thioesterase 6c14_5530, C14orf42, chromosome 14 open reading frame 42, EC 3.1.2.-, EC 3.1.2.2, MGC161656, MGC161658, putative acyl-coenzyme A thioesterase 6 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 641372 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction