missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACOT7 Antibody (1D5), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22569G
This item is not returnable.
View return policy
Description
ACOT7 Monoclonal antibody specifically detects ACOT7 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| ACOT7 | |
| Monoclonal | |
| DyLight 488 | |
| ACH1, acyl-CoA thioesterase 7CTE-IIa, BACHEC 3.1.2.2, brain acyl CoA hydrolase, Brain acyl-CoA hydrolase, CTE-IILACH, cytosolic acyl coenzyme A thioester hydrolase, hBACH, long chain, Long chain acyl-CoA thioester hydrolase | |
| Recombinant human ACOT7 (1-370aa) purified from E. coli | |
| 0.1 mL | |
| Neuroscience, Signal Transduction | |
| 11332 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
| 1D5 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction