missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACOX1 Antibody (102), HRP , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89866H
This item is not returnable.
View return policy
Description
ACOX1 Monoclonal antibody specifically detects ACOX1 in Human samples. It is validated for ELISA
Specifications
| ACOX1 | |
| Monoclonal | |
| HRP | |
| ACOXEC 1.3.3.6, acyl-CoA oxidase 1, palmitoyl, acyl-CoA oxidase, straight-chain, acyl-Coenzyme A oxidase 1, palmitoyl, AOX, PALMCOX, Palmitoyl-CoA oxidase, peroxisomal acyl-coenzyme A oxidase 1, peroxisomal fatty acyl-CoA oxidase, SCOXMGC1198, Straight-chain acyl-CoA oxidase | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human ACOX1 (AAH08767.1; Met1-Leu660). | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 51 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 102 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction