missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22214-1-AP-150UL
This item is not returnable.
View return policy
Description
Low molecular weight protein tyrosine phosphtases, including the human red cell phosphatase, HCPTP, are widely expressed cytosolic proteins of approximately 18kDa that exist in distinct isoforms and are highly selective for phosphotyrosine over phosphoserine or phosphotyrosine. The HCPTP-A and HCPTP-B proteins expressed in human red cells and placenta are fast and slow forms of red cell acid phosphatase5. Like other tyrosine phosphatases, HCPTP is sensitive to inhibition by vanadate, and has a catalytic mechanism that involves formation of a cystinylphosphate intermediate6. The crystal structure of the bovine heart derived enzyme BHPTP shows a four-stranded central parallel b sheet with flanking a helices and an active site cysteine residue in the typical tyrosine phosphatase sequence context, CXXXXXR.Specifications
| ACP1 | |
| Polyclonal | |
| Unconjugated | |
| ACP1 | |
| acid phosphatase 1, soluble, ACP1, Adipocyte acid phosphatase, HAAP, LMW PTP, LMW PTPase, Red cell acid phosphatase 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11431, 52 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P24666, Q9D358 | |
| ACP1 | |
| ACP1 Fusion Protein Ag17539 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction