Learn More
Waters Corporation ACQUITY HSS T3 Column
Product Code. 13149929
Encompass
Click to view available options
Length (Metric):
50 mm
75 mm
100 mm
150 mm
Diameter (Metric):
1 mm
2.1 mm
3 mm
Unit Size:
Each
Pack of 3
Product Code. 13149929
Brand: Waters Corporation 176001129
The ACQUITY HSS T3 UPLC columns exhibit ultra-low MS bleed and are fully scalable and transferable to HPLC separations.
  • The universal, silica-based bonded phase used for the ACQUITY HSS T3 sorbents is compatible with 100% aqueous mobile phase and should be your first choice when developing separations for polar and non-polar compounds.
Specifications
Diameter (Metric) 1 mm
Length (Metric) 100 mm
Max. Pressure 18000 psi (1240 Bar)
Particle Shape Spherical
Particle Size 1.8 μm
Pore Size 100 Å
Quantity 3/Pk.
USP Type L1
Endcapped Yes
Type Column