Learn More
Waters Corporation ACQUITY UPLC BEH C18 Column
As the premier column for method development, the ACQUITY UPLC BEH C18 sorbent provides you with the most universal column choice for UPLC separations.
Brand: Waters Corporation 186002346
Additional Details : Weight : 0.50000kg
Description
- The trifunctionally bonded BEH particle gives you the widest usable pH range (pH 1-12), superior low pH stability, and ultra-low column bleed for high sensitivity MS applications.
Specifications
1 mm | |
18000 psi (1240 Bar) | |
1.7 μm | |
1/Ea. | |
Yes |
100 mm | |
Spherical | |
130 Å | |
L1 | |
Column |