Learn More
Waters Corporation ACQUITY UPLC BEH C18 Column
As the premier column for method development, the ACQUITY UPLC BEH C18 sorbent provides you with the most universal column choice for UPLC separations.
€ 1410.00 - € 4545.00
Specifications
Max. Pressure | 18000 psi (1240 Bar) |
---|---|
Particle Shape | Spherical |
Particle Size | 1.7 μm |
Pore Size | 130 Å |
USP Type | L1 |
Product Code | Brand | Diameter (Metric) | Length (Metric) | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Length (Metric) | Quantity | Price | Quantity & Availability | |||
10456754
|
Waters Corporation
186002350 |
2.1 mm | 50 mm | 1/Ea. |
€ 1447.50
Each |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||
10075513
|
Waters Corporation
186002352 |
2.1 mm | 100 mm | 1/Ea. |
€ 1530.00
Each |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||
13147569
|
Waters Corporation
176000862 |
1 mm | 100 mm | 3/Pk. |
€ 4290.00
Pack of 3 |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||
10516053
|
Waters Corporation
176000863 |
2.1 mm | 50 mm | 3/Pk. |
€ 4035.00
Pack of 3 |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||
10240505
|
Waters Corporation
176000864 |
2.1 mm | 100 mm | 3/Pk. |
€ 4290.00
Pack of 3 |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||
13260250
|
Waters Corporation
186002346 |
1 mm | 100 mm | 1/Ea. |
€ 1530.00
Each |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||
10335954
|
Waters Corporation
186002349 |
2.1 mm | 30 mm | 1/Ea. |
€ 1410.00
Each |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||
13280250
|
Waters Corporation
186002353 |
2.1 mm | 150 mm | 1/Ea. |
€ 1635.00
Each |
Estimated Shipment: 02-05-2024
Log in to see stock. |
|||
13137569
|
Waters Corporation
176000861 |
1 mm | 50 mm | 3/Pk. |
€ 4035.00
Pack of 3 |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||
13139909
|
Waters Corporation
176001044 |
1 mm | 150 mm | 3/Pk. |
€ 4545.00
Pack of 3 |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||
13179909
|
Waters Corporation
176001048 |
2.1 mm | 150 mm | 3/Pk. |
€ 4545.00
Pack of 3 |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||
13169939
|
Waters Corporation
176001304 |
2.1 mm | 30 mm | 3/Pk. |
€ 3930.00
Pack of 3 |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||
10145835
|
Waters Corporation
186002344 |
1 mm | 50 mm | 1/Ea. |
€ 1447.50
Each |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||
13270250
|
Waters Corporation
186002347 |
1 mm | 150 mm | 1/Ea. |
€ 1635.00
Each |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||
13293830
|
Waters Corporation
186004690 |
3 mm | 150 mm | 1/Ea. |
€ 1635.00
Each |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||
13223810
|
Waters Corporation
186004660 |
3 mm | 50 mm | 1/Ea. |
€ 1447.50
Each |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||
13233810
|
Waters Corporation
186004661 |
3 mm | 100 mm | 1/Ea. |
€ 1530.00
Each |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||
15382577
|
Waters Corporation
186005604 |
2.1 mm | 75 mm | 1/Ea. |
€ 1492.50
Each |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||
13230000
|
Waters Corporation
176001794 |
3 mm | 30 mm | 3/Pk. |
€ 3930.00
Pack of 3 |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||
13213810
|
Waters Corporation
186004659 |
3 mm | 30 mm | 1/Ea. |
€ 1410.00
Each |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||
13250000
|
Waters Corporation
176001796 |
3 mm | 100 mm | 3/Pk. |
€ 4290.00
Pack of 3 |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||
13240000
|
Waters Corporation
176001795 |
3 mm | 50 mm | 3/Pk. |
€ 4035.00
Pack of 3 |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||
13260000
|
Waters Corporation
176001797 |
3 mm | 150 mm | 3/Pk. |
€ 4545.00
Pack of 3 |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||
17123212
|
Waters Corporation
186005609 |
3 mm | 75 mm | 1/Ea. |
€ 1492.50
Each |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||
Description
- The trifunctionally bonded BEH particle gives you the widest usable pH range (pH 1-12), superior low pH stability, and ultra-low column bleed for high sensitivity MS applications.
Specifications
18000 psi (1240 Bar) | |
1.7 μm | |
L1 | |
Column |
Spherical | |
130 Å | |
Yes |