Learn More
Invitrogen™ Acrolein Monoclonal Antibody (10A10), FITC
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA545766
Description
A 1:1,000 dilution of MA5-45766 was sufficient for detection of Acrolein in 2 μg of Acrolein conjugated to BSA by ECL immunoblot analysis using Goat Anti-Mouse IgG:HRP as the secondary Antibody. Specific for Acrolein modified proteins. Does not detect free acrolein. Does not cross-react with Crotonaldehyde, Hexanoyl Lysine, 4-Hydroxy-2-hexenal, 4-Hydroxy nonenal, Malondialdehyde, or Methylglyoxal modified proteins.
Acrolein is primarily used as an intermediate in the synthesis of acrylic acid and as a biocide. Acrolein may be formed from the breakdown of certain pollutants in outdoor air or from the burning of organic matter including tobacco, or fuels such as gasoline or oil. Acrolein is toxic to humans following inhalation, oral or dermal exposures.
Specifications
| Acrolein | |
| Monoclonal | |
| 1 mg/mL | |
| PBS, 9.1mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative; pH 7.4 | |
| Mouse | |
| Protein G | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 10A10 | |
| FITC | |
| 2-propenal; Acraldehyde; Acrolein; Acrylic aldehyde | |
| Synthetic Acrolein modified Keyhole Limpet Hemocyanin (KLH). | |
| 100 μg | |
| Primary | |
| 4°C, store in dark | |
| Liquid | |
| Chemical |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.