Learn More
Thermo Scientific™ AcroMetrix™ HBV Controls
Product Code. 18816054 Shop All Thermo Scientific Products
Change view
Click to view available options
Level:
Low
Mid
Unit Size:
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Level Unit Size
18816054 Mid Pack of 5
18806054 Low Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18816054 Supplier Thermo Scientific™ Supplier No. 965002
 Restricted Item
Estimated Shipment: 04-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Identify increases in random or systematic error of nucleic acid tests for hepatitis B viral DNA (HBV DNA) with these controls, which are for In Vitro Diagnostic Use.
Identify increases in random or systematic error of nucleic acid test procedures for the quantitative and qualitative determination of hepatitis B viral DNA (HBV DNA) with the Thermo Scientific™ AcroMetrix™ HBV Controls. Routine use of external run controls enables laboratories to monitor day-to-day test variation, lot-to-lot performance of test kits, and operator variation and can assist in identifying increases in random or systematic error. The controls are for In Vitro Diagnostic Use.
TRUSTED_SUSTAINABILITY
Specifications
Description AcroMetrix HBV Mid Control
Detectable Analytes HBV
Type HBV Control
Level Mid
Packaging 5 x 1mL
Quantity 5 x 1 mL