Learn More
Cytiva (formerly Pall Lab) AcroPak™ 1500 Capsules with Supor™ EKV Membrane
AcroPak Capsules for buffers, tissue culture media, and other biological fluids.
Brand: Cytiva (formerly Pall Lab) 12467
Additional Details : Weight : 0.38950kg
Description
- Newer technology recommended for simplified scale-up
- High filtration area and compact size are ideal for upscale trials
- Designed for research applications only
- Gamma-irradiated
Specifications
Hydrophilic PES | |
Sterile | |
1/2 in. Hose Barb |
0.2 μm | |
1500 cm2 | |
1/Ea. |